Porcelain Evaporating Dish, Flat Form, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Shallow form flat bottom evaporating dish.
- Shallow-form porcelain evaporating dishes with lip and flat bottom
- Glazed inside and out, except for rim and part of outside bottom
- Autoclavable
- Withstand temperatures up to 1150 °C (2102 °F)
Pack of six shallow-form porcelain evaporating dishes with lip and flat bottom. Dishes are glazed inside and out, except for the rim and part of the outside bottom. Autoclavable and designed to withstand temperatures up to 1150 °C (2102 °F); temperature change rate should not exceed 200 °C (392 °F) per hour.
Customers Who Bought This Also Bought