Electrometer Accessories
# de catalogue 470346-480
Fournisseur: American 3B Scientific
Spécifications
- Description:Electrometer accessories
- Cat. no.:470346-480
Spécifications
A propos de cet article
Set of accessories in combination with the 3b American scientific electrometer.
- Set of accessories for carrying out basic experiments on electrostatics, electricity and the photoelectric effect
Set (in combination with the 3b American Scientific Electrometer) includes 1 Faraday cup; 1 pair of friction rods; 1 metal rod with 4 mm drilled hole; 1 safety adapted socket; 1 plug-in capacitor 1 nF; 1 plug-in capacitor 10 nF; 1 plug-in resistor 100 MΩ; 1 pug-in resistor 1 GΩ; 1 plug in resistor 10 GΩ; 1 zinc electrode; and 1 grid electrode. Electrometer is required, but sold separately.
Ordering information: Electrometer is required, but sold separately. See catalog item 470233-810.
Delivery: Includes a 12 VAC plug-in power supply.
Customers Who Bought This Also Bought