VWR® Aluminum Canisters and Stainless Steel Frames for Blood Bag Storage
Fournisseur: CRYSTAL TECHNOLOGY & INDUSTRIES
Certificats
A propos de cet article
Organize, maximize storage and protect sample integrity of blood bags, bone marrow, stem cells, cord blood and plasma bags with durable canisters and frames.
- Canisters designed to hold Baxter Fenwal, CharterMed, Gambro, Stericon, MedSep and origen blood bags
- Multiple canister capacity options fit your storage needs
- Canisters have locking handle and thumb hole for easy handling
- Frames constructed of durable corrosion resistant stainless steel
- Multiple frames options available to fit your storage requirements
- Frames feature handles for easy retrieval of canisters
Complete offering of canisters and frames to fit your storage requirements.
Packaging: All canisters and frames carry our lifetime warranty.
Customers Who Bought This Also Bought