ContactUsLinkComponent
Innovating Science® Properties of Polymers
Innovating Science® Properties of Polymers
# de catalogue 470343-436
Fournisseur:  Ward's Science
PROPERTIES OF POLYMERS
Innovating Science® Properties of Polymers
# de catalogue 470343-436
Fournisseur:  Ward's Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Properties of polymers
  • Cat. no.:
    470343-436
  • No. of Uses:
    15
Spécifications
A propos de cet article
Learn about polymer chemistry polymer properties like cross-linking and super-absorbance.
  • Students will perform 3 polymer activities - making polymer worms, making polymer slime, and making polymer snow.
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-2, MS-PS1-3, HS-PS1-2, HS-PS2-6
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include structure and function, patterns
  • Engineering practices include analyzing and interpreting data, obtaining evaluating and communicating information
Polymers affect every aspect of our daily lives. From the plastics we use to the clothes we wear;
Polymer chemistry is everywhere. In this kit, students will perform three fun and fascinating activities involving polymers. In the first two activities, students will perform a cross-linking procedure on polymers to create polymer 'worms' and the classic 'slime' In the third activity, students will investigate the properties of a super-absorbent polymer, capable of absorbing hundreds of times its own weight in water, to create instant polymer snow.The kit contains enough materials for 15 groups. Teacher's Manual and Student Study Guide copymasters are included.
Delivery: Kit includes instructional manual and SDS booklet.
Customers Who Bought This Also Bought