Bath Lids for Refrigerated/Heating Circulators, Julabo
Fournisseur: JULABO
Certificats
A propos de cet article
Condensation trap bath tank lid. Reduces/eliminates moisture condensation in bath fluid at <10 °C operation. Lid locks in place with fluid fill port and outlet for connection to nitrogen gas purge. Use with refrigeration models: 600F, 601F, 1000F, 1000FW.
Locking bath tank lid. Reduces fluid vapor during high temperature operation. Lid locks in place with fluid fill port. Use with refrigeration models: 600F, 601F, 1000F, 1000FW and heating bath BC12.
Delivery: Includes all tools and components for assembly / installation.
Packaging: Ships via ground.
Customers Who Bought This Also Bought