Wall Cabinets, Blickman
Fournisseur: BLICKMAN INDUSTRIES
Certificats
A propos de cet article
Stainless steel wall cabinets and sloped tops are constructed with quality 304 stainless steel, #4 polish and have a seamless welded face preventing dirt collection while providing long lasting durability through unparalleled strength and corrosion resistance.
- Available with 13" depth, 18³/₄ to 47" width and 30 to 48" height
- Designed for flush face construction with a recessed back panel for uneven wall surfaces
- Solid stainless steel doors are double walled for added strength and sound deadened for optimal performance
- Stainless steel and ¹/₄" tempered glass door construction
- Sliding doors ride quietly on noiseless ball bearing rollers
- Comes with two or three ¹/₂" adjustable stainless steel shelves
- Sloped top available in 18³/₄ to 47" widths, aids in keeping surgical area sterile and is easily affixed to cabinet tops by factory-aligned stainless steel fasteners
Wall cabinets are designed to stand alone with finished sides or to fit within a framework of cabinets. It is constructed to fit flush against wall surfaces with a recessed back panel for uneven walls.
BodyTextAccessories: Sloping tops are available for Blickman wall cabinets and help prevent dust accumulation, provide for ease-of-cleaning and aid in keeping the surgical area sterile. 304 stainless steel. Easily affixed to cabinet tops by factory-aligned, stainless steel fasteners.
Customers Who Bought This Also Bought