High Cabinets, Blickman
Fournisseur: BLICKMAN INDUSTRIES
Certificats
A propos de cet article
Stainless steel high cabinets and sloped tops are constructed with quality 304 stainless steel, #4 polish and have a seamless welded face preventing dirt collection while providing long lasting durability through unparalleled strength and corrosion resistance.
- Available with 84" height, 18³/₄ to 47" width, 18" and 23" depth
- Designed to stand alone or fit with a framework of cabinets
- Constructed as floor and wall supported individual cases with finished sides
- Solid stainless steel doors are double walled for added strength and sound deadened for optimal performance
- Stainless steel and ¹/₄" tempered glass door construction
- Sliding doors ride quietly on noiseless ball bearing rollers
- Each contain 5 adjustable stainless steel shelves
- Sloped top available in 18³/₄ to 47" widths, aids in keeping surgical area sterile and is easily affixed to cabinet tops by factory-aligned stainless steel fasteners
High cabinets are designed to stand alone or fit within a framework of cabinets. Constructed as floor and wall supported individual cases with finished sides.
BodyTextAccessories: Sloping tops are available for Blickman high cabinets and help prevent dust accumulation, provide for ease-of-cleaning and aid in keeping the surgical area sterile.
Customers Who Bought This Also Bought