PUREZERO* HM4 Pouch-Style Face Mask
# de catalogue 76558-122
Fournisseur: O&M HALYARD CANADA, INC CA
Spécifications
- Description:PUREZERO* HM4 Pouch-Style Face Mask
- Cat. no.:76558-122
Spécifications
A propos de cet article
The PUREZERO* HM4 Pouch Mask is a three-layer mask with nylon headband that provides a high particle, bacterial, and viral filtration efficiency, as well as an expanded breathing chamber.
- Three-layer mask
- Nylon headband
- For industrial and cleanroom use
- Recommended for ISO Class 4 and above applications
Masks feature an exclusive SO SOFT* biocomponent technology inner lining for comfort.
Customers Who Bought This Also Bought