Well Plate Tubes for Bond Elut NEXUS & Bond Elut NEXUS WCX Plates, Agilent Technologies | Avantor
Well Plate Tubes for Bond Elut NEXUS & Bond Elut NEXUS WCX Plates, Agilent Technologies
6 ml, NEXUS WCX, 150 mg
Catalog # CAAG75520030
Fournisseur #:  75520030
Well Plate Tubes for Bond Elut NEXUS & Bond Elut NEXUS WCX Plates, Agilent Technologies
Catalog # CAAG75520030
Well Plate Tubes for Bond Elut NEXUS & Bond Elut NEXUS WCX Plates, Agilent Technologies
6 ml, NEXUS WCX, 150 mg
Price
$581.00
Log in pour la disponibilité
Pack of 96
Product Details & Documents
Bond Elut NEXUS is ideal for extractions from highly viscous samples.
  • Large particle size allows excellent flow for viscous samples
  • Nonconditioning method saves time and improves throughput
  • NEXUS WCX offers enhanced selectivity for certain analytes, such as quaternary amine drugs
Bond Elut NEXUS is an ultraclean polymeric sorbent, which has bimodal porosity and a high surface area. NEXUS offers a nonpolar retention mechanism with no preconditioning required. The large particle size makes NEXUS ideal for extractions from highly viscous samples, such as horse urine.
Bond Elut NEXUS WCX is a weak cation-exchange sorbent that offers extra selectivity for analytes such as quaternary ammonium drugs and anabolic steroids.
Spécifications
  • Capacité:
    6 ml
  • Bed Weight:
    150 mg
  • Phase:
    NEXUS WCX