Well Plate Tubes for Bond Elut Certify II Plates, Agilent Technologies
# de catalogue CAAG7551801C
Fournisseur: AGILENT TECHNOLOGIES, INC (CSD) CA
Spécifications
- Volume:1.8 ml
- Description:VersaPlate well plate tubes, Certify II, 100 mg, PE frit (20 µm)
- Cat. no.:CAAG7551801C
Spécifications
A propos de cet article
Bond Elut Certify II is a SAX/nonpolar SPE phase for extracting acidic analytes from biomatrices.
- Ideal for nonpolar and anionic compounds
- Optimized for acidic drug analysis
- Bimodal, nonpolar and strong anion exchange
Bond Elut Certify II was developed specifically for the rapid extraction of acidic drugs and metabolites from urine and other biological matrices. Certify II is a mixed-mode cartridge packed with nonpolar C8 and SAX sorbent. It has been optimized for acidic drugs and other compounds that possess both nonpolar and anionic characteristics.
Customers Who Bought This Also Bought