Dissolution Verification Tools, Agilent Technologies
Fournisseur: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificats
A propos de cet article
A variety of different height gauges and spacers to meet all needed regulatory specifications.
- Available for the Agilent 708-DS and legacy instruments
Verification and calibration tools can help you to install your accessories properly. Agilent provides a variety of different height gauges and spacers to meet all needed regulatory specifications, such as mechanical qualification and physical parameter verification, for your dissolution test.
Customers Who Bought This Also Bought