Level IV Isolation Gown, Mortech
Supplier: Mortech Manufacturing
Certificates
About this item
These level 4 isolation gowns offer the highest level of protection available and are used for high risk circumstances. The lightweight, breathable design allows for all-day comfort. They offer pathogen resistance, protection against non-airborne diseases, and are an effective barrier for fluids.
- Level 4 Isolation
- 75% SMS 25% PE
- Blue
Customers Who Bought This Also Bought