Face Mask, KN95, Mortech
# de catalogue 76549-678
Fournisseur: Mortech Manufacturing
Spécifications
- Description:Face mask, KN95, 5 per box
- Size:8×6×2"
- Cat. no.:76549-678
Spécifications
A propos de cet article
Professional protection, disposable medical mask.
- White
- General use, non-surgical
Nonsterile. Filtration efficiency 95%. Flexible elastic ear tape. Multilayer filtration.
Certifications: FDA and CE approved.
Customers Who Bought This Also Bought