ContactUsLinkComponent
Replacement Cast Vacuum Filter For Stryker 986 Cast Vac, Mortech
Replacement Cast Vacuum Filter For Stryker 986 Cast Vac, Mortech
# de catalogue 76549-628
Fournisseur:  Mortech Manufacturing
VACUUM FILTER REPLACEMENT FOR CAST
Replacement Cast Vacuum Filter For Stryker 986 Cast Vac, Mortech
# de catalogue 76549-628
Fournisseur:  Mortech Manufacturing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Replacement cast vacuum filter for Stryker 986 cast vac
  • Cat. no.:
    76549-628
Spécifications
A propos de cet article
The BD986-500 vacuum filter is a brand new direct replacement for the Stryker 986-500 Cast Saw Vac filter. It can be used in both the Styker 986 Mobile and 987 Wall Mount vacuums. Replacing the filter every 6 months more more is recommended depending on usage.