Limb Lock, Mortech | Avantor
Limb Lock, Mortech
20 cm (8.25") width
Catalog # 76549-596
Supplier:   Mopec
Fournisseur #:  BC012
Limb Lock, Mortech
Catalog # 76549-596
Supplier:  Mopec
Limb Lock, Mortech
Limb Lock, Mortech
20 cm (8.25") width
Price
$905.39
Log in pour la disponibilité
Each
Product Details & Documents
Exclusively offered by Mopec, this simple self-adjusting design provides a safe and steady method to secure all limb specimens during any autopsy or pathology dissection.
  • Heavy gauge stainless steel
The Limb Lock’s unique notched V-shape offers all the benefits of a second set of hands. Multiple units can be utilized simultaneously for larger amputation specimens, and the unit can be easily cleaned with all standard disinfectants.
Spécifications
  • Taille:
    20 cm (8.25") width