Hexagonal Mass Set of 5, Plated
# de catalogue 470339-280
Fournisseur: United Scientific Supplies
Spécifications
- Description:Hexagonal mass set of 5, plated
- Cat. no.:470339-280
Spécifications
A propos de cet article
Zinc plated hexagonal masses.
- Set of five zinc-plated hexagonal masses
- Accurate and non-rolling
- Packaged in a sturdy cardboard box
- Suitable for ages 11+
Set of five zinc-plated hexagonal masses supplied in a sturdy cardboard box. Includes one each of 100 g, 500 g, and 1000 g, and two each of 200 g masses.
Customers Who Bought This Also Bought