Slotted Weight Set of 10, with Rack
Catalog # 470338-918
Supplier: United Scientific Supplies
Specifications
- Description:Slotted weight set of 10, with rack
- Cat. no.:470338-918
Specifications
About this item
Slotted brass weights for use with hangers.
- Set of 10 slotted brass weights
- Durable rack also included
- Also available as a set of 8 (WHST08) or 12 (WHST12)
Set of 10 high-quality slotted brass weights with durable rack.
Weights included: 1×500 g; 5×100 g; 1×50 g; 2×20 g; 1×10 g
Customers Who Bought This Also Bought