Deluxe Slotted Weight Set of 8, with Rack
# de catalogue 470339-624
Fournisseur: United Scientific Supplies
Spécifications
- Description:Deluxe slotted weight set of 8, with rack
- Cat. no.:470339-624
Spécifications
A propos de cet article
Slotted brass weights for use with hangers.
- Set of 8 slotted brass weights
- Durable rack also included
- Also available as a set of 10 (WHST10) or 12 (WHST12)
Set of 8 high-quality slotted brass weights with durable rack.
Weights included: 1×500 g; 2× 200 g; 1×100 g; 1×50 g; 2×20 g; 1×10 g
Customers Who Bought This Also Bought