Mapping Compass
# de catalogue 470340-254
Fournisseur: United Scientific Supplies
Spécifications
- Description:Mapping compass
- Cat. no.:470340-254
Spécifications
A propos de cet article
Easy to use folding mapping compass.
- Sight finder
- Markings in both metric and English scales
- Map scale ratio of 1:25000
Explore the basics of navigation and orienteering with this easy-to-use compass. Product features a sight finder, as well as markings in both metric and English scales. A detachable lanyard is also included. Map scale is displayed in mm with a ratio of 1:25000.
Customers Who Bought This Also Bought