Insect Net
# de catalogue 470339-646
Fournisseur: United Scientific Supplies
Spécifications
- Description:Insect net
- Cat. no.:470339-646
Spécifications
A propos de cet article
Sturdy aluminum and nylon insect net.
- Nylon net
- Aluminum frame
- 14" diameter ring
This quality built nylon insect net features a telescoping aluminum handle that extends from 18 to 32". A simple twist of the handle locks and unlocks the handle for varying lengths. A 48" long conical net is mounted on a 14" ring.
Customers Who Bought This Also Bought