ContactUsLinkComponent
Porcelain Spatula
Porcelain Spatula
# de catalogue 470339-284
SPATULA PORCELAIN EA
Porcelain Spatula
# de catalogue 470339-284
Spécifications
  • Description:
    Porcelain spatula, 8" long
  • Cat. no.:
    470339-284
Spécifications
A propos de cet article
Easy grip glazed porcelain spatula.
  • Knobbed handle for easy grip
  • Measures 8" (20 cm) long
  • Autoclavable
This glazed porcelain laboratory spatula is designed with one long, flat end and a knobbed handle for a secure grip. Tool measures 8" long.
Customers Who Bought This Also Bought