ContactUsLinkComponent
Linearity FD Cardiac Markers for PATHFAST Immunoanalyzer, AUDIT MircoControls
Catalogue # 76540-916
Fournisseur:  Audit Microcontrols
Numéro CAS:  
undefined
Linearity FD Cardiac Markers for PATHFAST Immunoanalyzer, AUDIT MircoControls
Catalogue # 76540-916
Fournisseur:  Audit Microcontrols
Supplier Number:  K982M-5
Numéro CAS:  
Spécifications
  • Description:
    Linearity FD cardiac markers for PATHFAST immunoanalyzer
  • Size:
    1 ml
  • Cat. no.:
    76540-916
Spécifications
À propos de cet article
The linearity FD cardiac markers for PATHFAST immunoanalyzer is intended to simulate human patient serum samples for purpose of determining linearity, calibration verification and verification of reportable range for NTproBNP and troponin I.