VWR® Cleanroom 16" Nitrile Sterile Gloves
Supplier: VWR International
Certificates
About this item
VWR® cleanroom soft, nitrile gloves are manufactured from carboxylated acrylonitrile butadiene that provides reliable, durable and comfortable hand protection during demanding cleanroom applications.
- Designed for elbow length protection
- Ambidextrous, designed with micro textured fingers and palm, beaded cuff
- Not made from natural rubber latex
- Static-dissipative
- Single-use only
Cleanroom gloves, nitrile, sterile, GRADE A, 400 mm.
Specifications
- Catalog No:
- 76557-808
- 76557-810
- 76557-800
- 76557-802
- 76557-804
- 76557-806
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Cuff length:
- 16"
- 16"
- 16"
- 16"
- 16"
- 16"
- Cuff thickness:
- 4 mil
- 4 mil
- 4 mil
- 4 mil
- 4 mil
- 4 mil
- Finger thickness:
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- Palm thickness:
- 5 mil
- 5 mil
- 5 mil
- 5 mil
- 5 mil
- 5 mil
- Tensile strength:
- >18 MPa
- >18 MPa
- >18 MPa
- >18 MPa
- >18 MPa
- >18 MPa
- Ultimate elongation:
- 500%
- 500%
- 500%
- 500%
- 500%
- 500%
- Anti-static:
- +
- +
- +
- +
- +
- +