VWR® Cleanroom 16" Nitrile Non Sterile Gloves
Fournisseur: VWR International
Certificats
A propos de cet article
VWR® cleanroom soft, nitrile gloves are manufactured from carboxylated acrylonitrile butadiene that provides reliable, durable and comfortable hand protection during demanding cleanroom applications.
- Designed for elbow length protection
- Ambidextrous, designed with micro textured fingers and palm, beaded cuff
- Not made from natural rubber latex
- Static-dissipative
- Single-use only
Cleanroom gloves, nitrile, non sterile, ISO 4, 400 mm, VWR®.
Spécifications
- Catalog No:
- 76557-788
- 76557-790
- 76557-792
- 76557-794
- 76557-796
- 76557-798
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Finger thickness:
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- Anti-static:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes