Exhaust Filters V2.0, SCAT
# de catalogue 76470-488
Fournisseur: S.C.A.T.
Spécifications
- Description:Exhaust filter M, V2.0, with splash guard and change indicator, service life 6 months
- Cat. no.:76470-488
Spécifications
A propos de cet article
The SCAT Exhaust Air Filters V2.0 made of durable HD-PE material are high-performance exhaust air filters of the latest generation for use with SafetyWasteCaps or the Universal Waste Hubs "Jan. With its specific surface area of 1200 m²/g, the multi-component granulate is the optimal filter medium for almost all solvent vapors. Thanks to the optimized particle size of the activated carbon, escaping gases or vapors from waste disposal containers are reliably cleaned and converted back into healthy breathing air.
- Available in sizes (S/M/L) with different service lives (3/6/12 months)
- Available as both single pack and economy pack
- Filled with activated carbon optimized for solvent vapors and VOCs
- The pore structure of the activated carbon prevents sticking or clogging
- Over 99% of volatile substances are captured and neutralized
- Precise indication of the next replacement time by means of an indicator
- The use of exhaust filters can contribute to the reduction of the air exchange rate
Each economy pack is designed with exhaust air filters for a service life of 12 months and offers attractive cost advantages compared to individual purchases.
For further details, please refer to the Related Information section below.
Customers Who Bought This Also Bought