VWR® Salmon Cleanroom Paper
Supplier: VWR International
Certificates
About this item
Latex-Free paper has been injected with a synthetic copolymer to substantially reduce particle generation.
- Exceptional thickness and strength, with excellent heat resistance
- High opacity and 'class 10 certified'
Ideal for use in printers and high speed photocopies.
Specifications
- Catalog No:
- 76547-894
- 76547-912
- 76547-910
- Environmentally Preferable:
- Sustainable Materials, Waste Reducing
- Sustainable Materials, Waste Reducing
- Sustainable Materials, Waste Reducing