Masterflex® Single Use Tubing Pinch Clamps, Avantor®
Supplier: VWR International
Certificates
About this item
Clamps are double bagged and ready for use in your cleanroom environment.
- Sturdy glass reinforced nylon design
- Validated to USP Class VI
- Rated to 100 PSI (6,5 bar)
- Feature anti-tamper zip-tie lock out
- Sterilisable via autoclave or gamma irradiation (25 to 43 kG)
- Features removable sled for locking clamp in place
- Injection molded and assembled in ISO cleanroom
These clamps are manufactured, assembled, and packaged within an ISO cleanroom.
Specifications
- Catalog No:
- MFLX19999-97
- MFLX19999-96
- Max. pressure:
- 100 PSI
- 100 PSI
- Sterilisation:
- Autoclave, gamma
- Autoclave, gamma
- Regulatory approvals:
- USP Class VI
- USP Class VI