Hand-Operated Pump Dispensers for Barrels and Canisters
Fournisseur: BURKLE INC
Certificats
A propos de cet article
Ideal for contamination-free removal of organic solvents.
- Designed for use with 15.8 gal. (60 L) containers
- Sturdy stainless steel and PTFE construction ensures long life
- Suitable for containers with R 2” thread or pull-out spout
The hand-operated solvent pumps, with a capacity of 10 L per minute, safely dispense organic solvents and flavoring agents. Two ball valves prevent dangerous vapors from escaping after dispensing by providing a gas-tight, completely sealable pump. Model 06043-91 is ideal for stainless steel barrels with a R 2” thread, while model 06043-93 is suitable for tin-foil canisters with a pull-out spout. Order optional antistatic grounding set for pumping flammable liquids.
Delivery: Includes thread adapter for 06043-93.
Spécifications
- Catalog No:
- MFLX06043-93
- Tube length, inlet:
- 60 cm (23.625")
- Wetted materials:
- 304 stainless steel, PTFE
- Warranty:
- 15 months