Foot-Operated Pump Dispensers for Barrels, Canisters, and Tanks
Fournisseur: BURKLE INC
Certificats
A propos de cet article
Keep hands-free for safe dispensing.
- Large-volume foot-operated bellows for high pumping capacity
- Universal bung constructed of soft PVC for 1-3/16" to 2-3/4" (30 to 70 mm) openings
- Telescopic immersion tube adapts itself to a variety of containers
- Different materials and styles suit a wide range of applications
Safely transfer liquids keeping both hands on the container you are filling. Choose from polypropylene (PP), PVDF, or 304 stainless steel (SS) wetted materials and dispensing by spout or hose.
Spécifications
- Catalog No:
- MFLX06040-33
- MFLX06040-35
- MFLX06040-39
- Outlet connection:
- Spout
- Spout
- Spout
- Tube ext.Ø, inlet:
- 1.8 cm (0.71")
- 2.2 cm (0.866")
- 1.6 cm (0.63")
- Tube length, inlet:
- 70 cm (27.5")
- 90 cm (35.5")
- 70 cm (27.5")
- Wetted materials:
- Polypropylene (PP)
- Polypropylene (PP)
- PVDF
- Warranty:
- 15 months
- 15 months
- 15 months