Bürkle Foot-Operated Barrel Pumps for Solvents
Supplier: BURKLE INC
Certificates
About this item
Safely transfer corrosive solvents.
- Stainless steel and PTFE construction withstands long-term exposure to aggressive solvents
- Foot-operated bellows leaves both hands free for more stable container filling
- Telescopic immersion tube adapts itself to a variety of container heights
- Fits a 2" thread creating a closed system to contain harmful vapors
Use this foot-operated bellows pump to safely dispense solvents from drums up to 55 gal. (220 L). Choose between a pump with a spout or a discharge hose; both models include a stopcock to contain vapors and prevent dripping. Order optional antistatic grounding set for pumping flammable liquids.
Specifications
- Catalog No:
- MFLX06043-94
- Outlet connection:
- Discharge hose with stopcock
- Tube ext.Ø, inlet:
- 32 cm (1.25")
- Tube length, inlet:
- 95 cm (37.5")
- Wetted materials:
- 304 SS, PTFE
- Warranty:
- 15 months