Value Plastics Sanitary Adapters, Barb to Sanitary Clamp, PVDF
Fournisseur: Avantor Fluid Handling
Certificats
A propos de cet article
Create a secure connection in your sanitary fitting system.
- Constructed of durable PVDF
These adapters are perfect to transition from sanitary clamp connections to a barb for flexible tubing. All adapters feature a flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow without particulate build-up. There are no sharp edges in the flow path to make cleaning very easy. These adapters offer good chemical resistance and may be sterilized by gamma irradiation, EtOH, or autoclave. Material meets the requirements for USP Class VI, ADCF, FDA, and are BPA-free.
Spécifications
- Catalog No:
- MFLX40617-54BLK
- MFLX40617-53BLK
- MFLX31513-62
- MFLX31513-61
- MFLX40617-59BLK
- MFLX40617-52BLK
- MFLX40617-80BLK
- MFLX31513-66
- MFLX40617-61BLK
- Sterilisation:
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Warranty:
- Shelf life
Spécifications
Customers Who Bought This Also Bought