Gowning Room Bootie and Shoe Rack, Stainless Steel, Metro
Supplier: Metro
Certificates
About this item
The ideal solution to store cleanroom booties or shoes.
- Stainless steel Super Erecta shelving combined with rods and tabs allows you to create customized compartments to fit your needs
- All type 304 stainless steel
- Three sizes available or configure to fit your needs
Laboratory and cleanroom environments can be challenging. The rigorous cleaning aspect of the gowning room means that special considerations must be made when putting the space together. For your gowning room needs, trust Metro's solution for shoe and bootie storage. With years of experience in the field, we know how to make the most of every lab space.
Customers Who Bought This Also Bought