Utility Carts, Cleanroom Grade, Stainless Steel, Metro
Supplier: Metro
Certificates
About this item
Cleanroom grade all stainless-steel utility cart.
- 5" polyurethane cart-washable 2 swivel/2 break stem casters
- All type 304 stainless steel shelves and posts
- Specifically designed to handle regular cleaning and sterilization
When your environment calls for premium solutions, every storage or transport product must work to preserve the contaminant-free conditions. Each piece of Metro’s Grade A Pharma cart down to the casters is specifically designed to handle regular cleaning and sterilization without compromising its structural integrity.
Customers Who Bought This Also Bought