Hedland Flowmeter Accessories
Supplier: BADGER METER INC
Certificates
About this item
Make the most of your Hedland flowmeters with these replacement parts and other accessories.
- Optimize performance of your flowmeter with these accessories
Replacement switches and cables are suitable for use with Hedland spring-loaded in-line flowmeters or turbine flowmeters.