PrimeLock® Benchtop Installation Tool, 1/4" to 1" OD tubing
# de catalogue MFLX30493-25
Fournisseur: ANDON SPECIALTIES INC.
Spécifications
- Description:Benchtop Installation Tool, 1/4" to 1" OD tubing
- Warranty:2 months
- Compatible with:PrimeLock High-purity PFA flare fittings, ¹/₄ - 1" ext.Ø tubing
- Cat. no.:MFLX30493-25
Spécifications
A propos de cet article
Easily install your High-Purity PFA Fittings.
- Optimize performance of your PFA PrimeLock® Fittings
Handheld and benchtop installation tools help you to quickly and easily install the PrimeLock insert correctly every time. Benchtop tool is the quickest and easiest method of installing the insert when space constraints are not an issue. The compact, handheld tool enables you to install the insert in remote locations. Installation tools require mandrel(s) and clamp insert(s) for operation.
- Description:Benchtop Installation Tool, 1/4" to 1" OD tubing
- Warranty:2 months
- Compatible with:PrimeLock High-purity PFA flare fittings, ¹/₄ - 1" ext.Ø tubing
- Cat. no.:MFLX30493-25
Customers Who Bought This Also Bought