Barblock Fitting and Tubing Tools and Accessories
Supplier: ST. GOBAIN- TRAVERSE CITY
Certificates
About this item
Easily install your Barblock Fittings.
- Optimize performance of your Barblock fittings
Select a hand tool to assemble your BarbLock Retainers based on the tubing size. Universal removal tool is also available.
Specifications
- Warranty:
- 6 months
- 6 months
- 6 months
- 6 months
- None