Harwil In-Line Liquid Level Switches
Fournisseur: VWR International
Certificats
A propos de cet article
Eliminate nuisance switch tripping with a wide range of differentials.
- Large differentials are immune to wave action and turbulence (L-21VCR models)
- Single-point switch provides pump up/pump down level control
- Withstands corrosive and particle-contaminated working fluids
This series of liquid level switches are easy to install, operate and maintain. Through-wall NPT process fittings and NPT electrical ports facilitate integration into existing systems. Operation checks and troubleshooting require only a multimeter.
Wetted materials are fiberglass-reinforced PPS and Hastelloy® C. Switch types and materials of construction resist corrosion and increase durability, making them suitable even for seawater and filtered sewage.
Spécifications
- Min. process temperature:
- 32 °F
- 32 °F
- 32 °F
- 32 °F
- 32 °F
- Max. process temperature:
- 93 °C
- 93 °C
- 93 °C
- 93 °C
- 93 °C
- Min. specific gravity:
- 0.7
- 0.7
- 0.8
- 0.7
- 0.7
- Max. pressure:
- 250 psi
- 250 psi
- 75 psi
- 250 psi
- 250 psi
- Relay output:
- 15 A at 125 or 250 VAC. 0.5 A at 125 VDC. 0.25 A at 250 VDC
- 15 A at 125 or 250 VAC. 0.5 A at 125 VDC. 0.25 A at 250 VDC
- 15 A at 125 or 250 VAC. 0.5 A at 125 VDC. 0.25 A at 250 VDC
- 15 A at 125 or 250 VAC. 0.5 A at 125 VDC. 0.25 A at 250 VDC
- 15 A at 125 or 250 VAC. 0.5 A at 125 VDC. 0.25 A at 250 VDC
- Wetted materials:
- Stem: Hastelloy C; Float: Fiberglass-reinforced PPS
- Stem: Hastelloy C; Float: Fiberglass-reinforced PPS
- Stem: Hastelloy C; Float: Fiberglass-reinforced PPS; Diaphragm: Viton
- Stem: Hastelloy C; Float: Fiberglass-reinforced PPS
- Stem: Hastelloy C; Float: Fiberglass-reinforced PPS
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year