Noshok 400 Series Precision Needle Valves
Fournisseur: VWR International
Certificats
A propos de cet article
Hard-seat design.
- Metal-to-metal hard seat design is 100% helium leak-tested to 1 x 10–4 ml/s at 200 psi
- 10,000 psi pressure rating at 200 °F (93 °C)
- Blowout-proof stem
Series 400 needle valves are high quality instrument grade valves. They are the right choice when your application calls for positive shut-off or precise flow control. They work effectively with most gases and liquids.
Spécifications
- Catalog No:
- MFLX06292-09
- MFLX06292-02
- Max. pressure:
- 10.000 psi
- 10.000 psi
- Orifice:
- 4.7498 mm (0.187")
- 11.1252 mm (0.438")
- Wetted materials:
- Stainless Steel
- Stainless Steel
- Warranty:
- 3 years
- 3 years