Masterflex® Smart Clamp® Stainless Steel Sanitary Clamps, Avantor®
Supplier: VWR International
Certificates
About this item
Torque control for consistent sealing of fittings.
- Constructed of durable stainless steel
These sanitary clamps feature a unique design that allows fast clamping while reducing the repetitive strain injuries in hands and fingers that can occur with regular sanitary clamps; only a half turn is required. Clamps also have an adjustable torque for compression control and feature multiple settings for use with a variety of sanitary gaskets. Threadless design prevents soil and bacteria entrapment. Clamps also support ASME-BPE standards.
Specifications
- Catalog No:
- MFLX61020-82
- MFLX31608-91
- Sanitary clamp size (port 1):
- 3"
- 2"
- Connection type:
- Sanitary
- Sanitary
- Warranty:
- None
- None