ContactUsLinkComponent
2"Threaded Buttress adapter for 06425-05
2" Threaded Buttress adapter for 06425-05
# de catalogue MFLX06425-09
2" Threaded Buttress adapter for 06425-05
2" Threaded Buttress adapter for 06425-05
# de catalogue MFLX06425-09
Spécifications
  • Connection type:
    Threaded Reducer
  • Description:
    2" Threaded Buttress adapter for 06425-05
  • Warranty:
    1 year
  • Fitting Material:
    HDPE
  • Flow Pattern:
    Straight
Spécifications
A propos de cet article
Optimize performance of your hand-operated pump.
  • Constructed of durable HPDE
This 2" threaded buttress adapter is made of HDPE and suitable for use with HDPE hand pump 06425-05.
  • Connection type:
    Threaded Reducer
  • Description:
    2" Threaded Buttress adapter for 06425-05
  • Warranty:
    1 year
  • Fitting Material:
    HDPE
  • Flow Pattern:
    Straight
Customers Who Bought This Also Bought