2" Threaded Buttress adapter for 06425-05
# de catalogue MFLX06425-09
Fournisseur: BECKSON INDUSTRIAL PRODUCTS
Spécifications
- Connection type:Threaded Reducer
- Description:2" Threaded Buttress adapter for 06425-05
- Warranty:1 year
- Fitting Material:HDPE
- Flow Pattern:Straight
Spécifications
A propos de cet article
Optimize performance of your hand-operated pump.
- Constructed of durable HPDE
This 2" threaded buttress adapter is made of HDPE and suitable for use with HDPE hand pump 06425-05.
- Connection type:Threaded Reducer
- Description:2" Threaded Buttress adapter for 06425-05
- Warranty:1 year
- Fitting Material:HDPE
- Flow Pattern:Straight
Customers Who Bought This Also Bought