TriClamp Sanitary Strainer Assemblies, 316 Stainless Steel
Fournisseur: RUBBERFAB GASKET & MOLDING
Certificats
A propos de cet article
Protect products and expensive process equipment from coarse or fine particulates.
- 316 SS fluid contact surfaces are polished to a smooth, blemish free finish
- Standard clamp sizes allow easy installation or retrofit
Sanitary strainers are ideal for particulate removal or pre-filtration in the processing of foods, beverages, pharmaceuticals, cosmetics, and other critical compounds. Housings are available in in-line and side-inlet configurations.
Delivery: Strainer assemblies include housing, perforated core, internal cap and spring, assembly clamp and gasket. Order wire mesh overscreens of the desired mesh size separately.
Spécifications
- Catalog No:
- MFLX29590-14
- MFLX29590-02
- Filter type:
- Strainer
- Strainer
- Length:
- 40.01 cm (15.75")
- 40.01 cm (15.75")
- Outlet connection:
- 2" Tri-Clamp®
- 1-1/2" Tri-Clamp®
- Warranty:
- None
- None