Parker Hannifin Ball Valves, Polypropylene
Fournisseur: KAMAN FLUID POWER
Certificats
A propos de cet article
Create a secure and easy push-to-connect connection.
- Glass reinforced polypropylene (PP) body with a polysulfone ball and EPDM seals
- NSF 51 listed
- FDA compliant for contact with food
These Parker Hannifin polypropylene (PP) ball valves feature an all plastic design is leak free and corrosion resistant.
Customers Who Bought This Also Bought