Hedland High TempFlow Meters for Petroleum and Water
Fournisseur: BADGER METER INC
Certificats
A propos de cet article
Direct read 360° rotatable scale.
- Easy-to-read linear scale
FEATURES
Direct readings
Install in any position
360° Rotatable Guard/Scale
No flow Straighteners or Special Piping Required
Insensitive to Shock and Vibration
Good Viscosity Stability
Temperature up to 500 °F
Accuracy +/- 2% Full Scale
Repeatability +/-1%
Special Scales Available
Spécifications
- Catalog No:
- MFLX32509-07
- MFLX32508-92
- Max. operating temperature:
- 260 °C (500 °F)
- 260 °C (500 °F)
- Warranty:
- 1 year
- 1 year