Great Plains Economy Fuel and Water Meters
Fournisseur: GREAT PLAINS INDUSTRIES INC
Certificats
A propos de cet article
Small, powerful, and durable.
- Battery powered 4 digit display
- Factory calibrated to water or fuel
- Gallon or litre measurement units
- Batch and cumulative totals
- Ready to install
Small, powerful, microprocessor based totalizers powered by AAA batteries have large four-digit displays and mount at the nozzle for ultimate convenience. They are suited for flow ranges from 3 to 30 GPM. Nylon® models, rated to 150 PSIG, are factory calibrated for water. Aluminum models, rated to 300 PSIG are factory calibrated for fuels. Accuracy is ±5%. All mount with 1 inch female NPT fittings.
Spécifications
- Catalog No:
- MFLX32486-02
- MFLX32486-01
- MFLX32485-99
- Wetted materials:
- Aluminum
- Nylon
- Aluminum
- Warranty:
- 1 year
- 1 year
- 1 year