GF Signet 3-8150-1 8150 Field Mount Flow Monitor/Totalizer, Battery Powered
# de catalogue MFLX32487-29
Fournisseur: BURT PROCESS EQUIPMENT
Spécifications
- Description:8150 Field Mount Flow Monitor/Totalizer, Battery Powered
- Warranty:1 year
- Cat. no.:MFLX32487-29
Spécifications
A propos de cet article
Easy read your flow.
- Loop-Powered Digital Flow Transmitter
- Battery-Powered Digital Flow Rate Monitor and Totalizer
- Line-Powered Analog/Digital Flow Controller
- Batch Controller
This field mount flow monitor/totalizer is battery powered for optimal convience.
- Description:8150 Field Mount Flow Monitor/Totalizer, Battery Powered
- Warranty:1 year
- Cat. no.:MFLX32487-29
Customers Who Bought This Also Bought