Parker High-Pressure Multimedia Solenoid Valves
Fournisseur: PARKER HANNIFIN PRECISION FLUIDICS
Certificats
A propos de cet article
Offer outstanding potential for precision control in liquid and gas analysis.
- High speed — typical response time is <5 ms
- Maximum pressure up to 1250 psi
- Constructed of noncorroding, passivated stainless steel
- Leak tight to 1 x 10-7 cc/sec/atm Helium
These rugged valves operate with high repeatability, combining high speed and high flow in a small size. Coils are rated continuous duty and are potted for protection from dust and dirt.
Spécifications
- Max. pressure:
- 86 bar (1250 psi)
- 17.2 bar (250 psi)
- 6.9 bar (100 psi)
- 6.9 bar (100 psi)
- Max. temperature:
- 105 °C (221 °F)
- 105 °C (221 °F)
- 105 °C (221 °F)
- 105 °C (221 °F)
- Media compatibility:
- gases and liquids compatible with wetted materials
- gases and liquids compatible with wetted materials
- gases and liquids compatible with wetted materials
- gases and liquids compatible with wetted materials
- Power:
- 24 VDC
- 24 VDC
- 24 VDC
- 24 VDC
- Response time:
- <5 ms
- <5 ms
- <5 ms
- <5 ms
- Seal material:
- Vespel®, FKM
- FKM
- FKM
- FKM
- Wetted materials:
- 316L SS, PTFE, and seals
- 316L SS, PTFE, and seals
- 316L SS, PTFE, and seals
- 316L SS, PTFE, and seals