Digital Ultrasonic Barrel and Drum Level Gauge
Fournisseur: VWR International
Certificats
A propos de cet article
Never come up empty—reads to ±0.50 gallon (1 liter).
- Resists temperature fluctuations
- Ideal for applications involving diesel fuel, oils, hydraulic fluid, and water†
- Works with 15, 30, or 55 gallon (57, 114, or 208 liter) vertical drums
- Digital display indicates volume in gallons or liters numerically and as an analog bar graph
Ultrasonic drum level gauges feature all laser-welded design and custom integrated signal conditioning for calibration and temperature compensation.
Spécifications
- Catalog No:
- MFLX68349-52
- MFLX68349-53
- Accuracy of depth range:
- 0.5% full scale
- ±2% full-scale volume
- Min. depth:
- 12.7 cm (5")
- 5"
- Max. depth:
- 102 cm (40")
- 102 cm (40")
- Min. process temperature:
- 32 °F
- 32 °F
- Max. process temperature:
- 50 °C
- 50 °C
- Wetted materials:
- 304/316 SS, glass-filled PBT, and HDPE
- 304/316 SS, glass-filled PBT, and HDPE
- Warranty:
- 1 year
- 1 year