Grooving Tool for PFA Compression Fittings
Grooving Tool for PFA Compression Fittings, 6 mm OD
Supplier: ANDON SPECIALTIES INC.
Fournisseur #: GT6M
Grooving Tool for PFA Compression Fittings
Catalog # MFLX31321-49
Supplier: ANDON SPECIALTIES INC.
CE certificate
Jump to:
Product Details & Documents
Get a secure seal at higher pressures.
- Optimize performance of your PFA Compression Fittings
Improve the sealing capability of PTFE fittings. Fittings withstand pressures to 150 psi when a tool is used (40 psi without tool).
Documents
Certificates
Recherche par numéro de lotSpécifications
- Désignation:Grooving Tool for PFA Compression Fittings, 6 mm OD
- Cat. No.:MFLX31321-49
- Warranty:1 year
- Compression size / tube ext.Ø (port 1):6 mm
- Connection type:Compression
- Sterilisation:Autoclave or ethylene oxide (ETO)