Parker Hannifin Union Fittings, Compression, Elbows
Fournisseur: KAMAN FLUID POWER
Certificats
A propos de cet article
Get a secure seal at higher pressures.
- Quick and easy assembly without tools
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Customers Who Bought This Also Bought