Multi-Channel Tubing Racks, Non-Adhesive
Fournisseur: VWR International
Certificats
A propos de cet article
An easy solution to secure and route multiple tubing lines.
- Keep your tubing lines organized and untangled
These tube racks offer a simple and economical method for organizing your tubing lines. The 3/16" and 1/4" sizes can be easily separated down to single holders by hand for increased flexibility and custom tubing arrangements. The 3/16" and 1/4" sizes are available with adhesive fastener, and all sizes are available with through holes for secure mounting (mounting screws are not included). Standard racks are black polyethylene; 3/16" and 1/4" sizes are available in other colors. Contact Cole-Parmer for more information.
Spécifications
- No. of channels:
- 10
- 7
- 5
- 10
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year